6-Quinolinecarboxaldehyde 97%

Supplier: Ambeed
Warning

A158138-10G A158138-250MG A158138-1G A158138-25G A158138-5G A158138-100G
103805-098EA 15 USD
103805-098 103843-306 76768-468 76768-470 77319-150 77319-152 77319-154 77877-045
6-Quinolinecarboxaldehyde 97%
6-Quinolinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
Boiling Pt: 314.3 °C (760 mm Hg)
Melting Pt: 74…75 °C
Flash Pt: 151.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00805836
CAS Number: 4113-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR