6-Phenyl-3(2H)-pyridazinone 97%

Supplier: Ambeed
Warning

Synonyms: 6-Phenylpyridazin-3(2H)-one, 3-Hydroxy-6-phenylpyridazine

A153000-1G A153000-5G
103804-456EA 15 USD
103804-456 103842-628 77318-262 77318-264
6-Phenyl-3(2H)-pyridazinone 97%
6-Phenyl-3(2H)-pyridazinone

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.19 g/mol
Boiling Pt: 403.8 °C (760 mm Hg)
Melting Pt: 201…204 °C
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00114953
CAS Number: 2166-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR