6-Phenylpicolinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Phenylpicolinic acid, 6-Phenylpyridine-2-carboxylic acid

A374742-1G A374742-5G A374742-10G A374742-250MG
103826-560EA 30 USD
103826-560 103837-578 76840-860 76840-862 76978-750 76978-752
6-Phenylpicolinic acid 95%
6-Phenylpicolinic acid

Precaución: For research use only

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
Boiling Pt: 409 °C (760 mm Hg)
Flash Pt: 201.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04114206
CAS Number: 39774-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR