6-Phenyl-3-formylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Phenyl-5-pyridinecarboxaldehyde, 6-Phenylnicotinaldehyde

A411868-100MG A411868-250MG A411868-1G A411868-5G
103827-230EA 57 USD
103827-230 103838-278 76817-978 76817-980 76841-818 76841-820
6-Phenyl-3-formylpyridine 95%
6-Phenyl-3-formylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₉NO
Boiling Pt: 340.4 °C (760 mm Hg)
Melting Pt: 57 °C at N/A mmHg
Flash Pt: 167.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03086130
CAS Number: 63056-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR