6-Hydroxynicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid

A297691-1000G A297691-10G A297691-500G A297691-100G
103835-292EA 5 USD
103835-292 77114-406 77114-408 77114-410 77593-802
6-Hydroxynicotinic acid 97%
6-Hydroxynicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 384 °C (760 mm Hg)
Melting Pt: 300 °C
Flash Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00006277
CAS Number: 5006-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR