6-Oxo-1,6-dihydropyrazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A170914-100MG A170914-250MG A170914-1G A170914-5G
103806-554EA 234 USD
103806-554 103844-830 76985-410 76985-412 77301-082 77301-084
6-Oxo-1,6-dihydropyrazine-2-carboxylic acid 95%
6-Oxo-1,6-dihydropyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697776
CAS Number: 13924-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR