6-Nitropicolinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Nitropyridine-2-carboxylic acid

A129353-100MG A129353-250MG A129353-1G A129353-5G
103801-268EA 43 USD
103801-268 76688-562 76688-564 77313-652 77313-654
6-Nitropicolinic acid 95%
6-Nitropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
Boiling Pt: 434.8 °C (760 mm Hg)
Melting Pt: 169…172 °C
Flash Pt: 203.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114205
CAS Number: 26893-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR