6-Nitropyridin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-nitropyridine

A159181-100MG A159181-250MG A159181-1G
103805-230EA 89 USD
103805-230 103843-446 77011-132 77011-134 77319-324
6-Nitropyridin-2-amine 98%
6-Nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₅N₃O₂
MW: 139.1121 g/mol
Melting Pt: 160…164 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114202
CAS Number: 14916-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR