2-Mercapto-6-nitrobenzothiazole 97%

Supplier: Ambeed
Warning

A212659-10G A212659-5G
103818-222EA 35 USD
103818-222 103848-918 76770-586 77087-388
2-Mercapto-6-nitrobenzothiazole 97%
2-Mercapto-6-nitrobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂S₂
Boiling Pt: 348.4 °C (760 mm Hg)
Melting Pt: 249…253 °C
Flash Pt: 164.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00041850
CAS Number: 4845-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR