6-Nitro-4-(trifluoromethyl)quinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A281637-250MG A281637-1G A281637-5G
103813-564EA 21 USD
103813-564 103853-512 76827-580 77305-410 77305-412
6-Nitro-4-(trifluoromethyl)quinolin-2(1H)-one 97%
6-Nitro-4-(trifluoromethyl)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₅F₃N₂O₃
Boiling Pt: 368.7 °C (760 mm Hg)
Melting Pt: 284…285 °C
Flash Pt: 176.8 °C
Storage Temperature: Ambient
MDL Number: MFCD12828492
CAS Number: 328956-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR