3,4-Dihydro-6-nitro-1H-quinolin-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 6-Nitro-3,4-dihydroquinolin-2(1H)-one

A324714-1G A324714-5G A324714-100G A324714-10G A324714-250MG
103825-378EA 18 USD
103825-378 103836-322 76839-240 76839-242 76875-932 76875-934 76875-936
3,4-Dihydro-6-nitro-1H-quinolin-2-one 95%
3,4-Dihydro-6-nitro-1H-quinolin-2-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MW: 192.171 g/mol
Boiling Pt: 420.3 °C (760 mm Hg)
Melting Pt: 201…202 °C
Storage Temperature: Freezer
MDL Number: MFCD00559317
CAS Number: 22246-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR