6-Nitroindole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Nitro-1H-indole
A151640-100G
A151640-10G
A151640-1G
A151640-25G
A151640-5G
103804-288EA
9
USD
103804-288
103842-440
76689-380
76689-382
77318-022
77318-024
77318-026
6-Nitroindole 98%
6-Nitroindole
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 362.6 °C (760 mm Hg) Melting Pt: 137…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051497 CAS Number: 4769-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...