5-Nitro-1H-benzo[d]imidazol-2-amine 97%

Supplier: Ambeed
Warning

A216681-5G A216681-10G A216681-250MG
103818-574EA 17 USD
103818-574 103849-296 77087-888 77139-770 77139-772
5-Nitro-1H-benzo[d]imidazol-2-amine 97%
5-Nitro-1H-benzo[d]imidazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₆N₄O₂
Boiling Pt: 478.5 °C (760 mm Hg)
Flash Pt: 243.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06659794
CAS Number: 6232-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR