6-Nitro-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

A124975-10G A124975-250MG
103800-600EA 39 USD
103800-600 76766-844 76766-846
6-Nitro-1,2,3,4-tetrahydroquinoline 95%
6-Nitro-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₂
Boiling Pt: 341.4 °C (760 mm Hg)
Flash Pt: 160.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00541472
CAS Number: 14026-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR