2-Hydroxy-6-methylbenzaldehyde 95%

Supplier: Ambeed
Warning

A157612-100MG A157612-10G A157612-250MG A157612-1G A157612-5G
103805-022EA 84 USD
103805-022 103843-224 76973-382 76973-384 76973-386 77300-270 77300-272
2-Hydroxy-6-methylbenzaldehyde 95%
2-Hydroxy-6-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
Boiling Pt: 217.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD16038308
CAS Number: 18362-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR