6-Methylquinolin-8-ol 98%

Supplier: Ambeed
Warning

A249584-100MG A249584-250MG A249584-1G
103811-882EA 221 USD
103811-882 103851-720 77013-954 77013-956 77304-496
6-Methylquinolin-8-ol 98%
6-Methylquinolin-8-ol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.18 g/mol
Boiling Pt: 328.1±22.0 °C (760 mm Hg)
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD18412519
CAS Number: 20984-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR