4-Hydroxy-6-methylquinoline 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methylquinolin-4-ol

A122877-100MG A122877-250MG A122877-5G
103800-298EA 29 USD
103800-298 77108-662 77108-664 77312-418
4-Hydroxy-6-methylquinoline 95%
4-Hydroxy-6-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.185 g/mol
Boiling Pt: 290.3 °C (760 mm Hg)
Melting Pt: 233…237 °C
Flash Pt: 131.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00272343
CAS Number: 23432-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR