6-Methyl-4-quinazolone 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methylquinazolin-4-ol, 6-Methylquinazolin-4(3H)-one

A122758-10G A122758-250MG A122758-1G A122758-5G
103800-278EA 29 USD
103800-278 77284-926 77284-928 77312-394 77312-396
6-Methyl-4-quinazolone 95%
6-Methyl-4-quinazolone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.1726 g/mol
Boiling Pt: 386.7±21.0 °C (760 mm Hg)
Melting Pt: 238…240 °C
Flash Pt: 156.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00673185
CAS Number: 19181-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR