6-Methylpyridine-3-sulfonic acid 98%

Supplier: Ambeed
Warning

A387906-1G A387906-100MG A387906-250MG
103826-834EA 51 USD
103826-834 103837-876 76841-240 77115-594 77115-596
6-Methylpyridine-3-sulfonic acid 98%
6-Methylpyridine-3-sulfonic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₃S
MW: 173.1897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646153
CAS Number: 4808-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR