6-Methylpyridine-2-thiol 97%

Supplier: Ambeed
Warning

A224350-250MG A224350-1G A224350-25G
103819-102EA 86 USD
103819-102 103849-992 77098-020 77303-692 77303-694
6-Methylpyridine-2-thiol 97%
6-Methylpyridine-2-thiol

Precaución: for Research Use Only

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 197.4 °C (760 mm Hg)
Melting Pt: 150…155 °C
Flash Pt: 73.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06411332
CAS Number: 18368-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR