5-Amino-2-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-picoline, 6-Methylpyridin-3-amine, 3Amino6methylpyridine, 6-Methyl-3-pyridinamine, 3Amino6picoline
A117855-100G
A117855-10G
A117855-25G
A117855-1G
A117855-5G
A117855-500G
103798-942EA
5
USD
103798-942
103824-834
77108-362
77108-364
77108-366
77311-614
77311-616
77685-042
5-Amino-2-methylpyridine 97%
5-Amino-2-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₈N₂ MW: 108.14 Boiling Pt: 238.4±20.0 °C (760 mm Hg) Melting Pt: 94…95 °C Flash Pt: 121.09 °C Storage Temperature: Ambient |
MDL Number:
MFCD00833389 CAS Number: 3430-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...