3-Amino-6-methylpyridazine 97%

Supplier: Ambeed
Warning

A351731-1G A351731-25G A351731-100G A351731-10G A351731-5G
103826-050EA 17 USD
103826-050 103837-046 76840-152 76840-154 77278-174 77278-176 77306-616
3-Amino-6-methylpyridazine 97%
3-Amino-6-methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 319 °C (760 mm Hg)
Melting Pt: 222…223 °C
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD01569899
CAS Number: 18591-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR