6-Methylpyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A489990-100MG A489990-10G A489990-250MG A489990-1G A489990-25G A489990-5G
103828-210EA 10 USD
103828-210 103839-282 76774-432 76774-434 76774-436 76843-240 76843-242 76843-244
6-Methylpyrazine-2-carboxylic acid 97%
6-Methylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
Boiling Pt: 302.8 °C (760 mm Hg)
Melting Pt: 138…140 °C
Flash Pt: 136.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08705756
CAS Number: 5521-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR