2-Amino-6-methylpyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methylpyrazin-2-amine

A234332-10G A234332-250MG
103810-940EA 17 USD
103810-940 103850-696 77261-602 77261-604
2-Amino-6-methylpyrazine 97%
2-Amino-6-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 246 °C (760 mm Hg)
Flash Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235193
CAS Number: 5521-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR