2-Cyano-6-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Methylpicolinonitrile, 6-Methyl-2-pyridinecarbonitrile, 6-Cyano-2-picoline, 6-Methylpyridine-2-carbonitrile
A156982-100G
A156982-10G
A156982-500G
A156982-1G
A156982-25G
A156982-5G
103804-950EA
5
USD
103804-950
103843-152
76812-484
76812-486
76812-488
77300-228
77318-960
77318-962
2-Cyano-6-methylpyridine 97%
2-Cyano-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₆N₂ MW: 118.14 g/mol Boiling Pt: 238.2 °C (760 mm Hg) Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190586 CAS Number: 1620-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...