6-Methylnicotinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid
A109326-100G
A109326-10G
A109326-500G
A109326-1G
A109326-5G
A109326-25G
103797-490EA
5
USD
103797-490
103823-286
77188-978
77188-980
77188-982
77283-858
77283-860
77614-836
6-Methylnicotinic acid 98%
6-Methylnicotinic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₂ Boiling Pt: 296.2 °C (760 mm Hg) Melting Pt: 210…213 °C Flash Pt: 132.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006341 CAS Number: 3222-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...