6-Methylcoumarin 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-2-benzopyrone, 6-Methyl-2H-chromen-2-one

A224181-100G A224181-10G A224181-500G A224181-5G A224181-25G A224181-1000G
103809-586EA 182 USD
103809-586 76770-950 76770-952 76770-954 76770-956 77088-622 77303-688
6-Methylcoumarin 97%
6-Methylcoumarin

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 304.6 °C (760 mm Hg)
Melting Pt: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00006875
CAS Number: 92-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR