6-Methyl-4-chromanone 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methylchroman-4-one

A347877-250MG A347877-1G A347877-5G
103825-980EA 22 USD
103825-980 103836-966 76694-072 77306-586 77306-588
6-Methyl-4-chromanone 95%
6-Methyl-4-chromanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 303.1 °C (760 mm Hg)
Melting Pt: 31…35 °C
Flash Pt: 145.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00024071
CAS Number: 39513-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR