6-Methyl-2-mercaptobenzothiazole 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methylbenzo[d]thiazole-2-thiol

A264396-1G A264396-5G A264396-250MG
103812-674EA 37 USD
103812-674 103852-566 77092-228 77092-230 77166-282
6-Methyl-2-mercaptobenzothiazole 95%
6-Methyl-2-mercaptobenzothiazole

Precaución: for Research Use Only

Formula: C₈H₇NS₂
MW: 181.278 g/mol
Boiling Pt: 322.6 °C (760 mm Hg)
Melting Pt: 183 °C
Flash Pt: 148.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07774732
CAS Number: 2268-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR