6-Methyl-2-pyridinemethanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxymethyl-6-methylpyridine, 6-Hydroxymethyl-2-picoline

A112772-100G A112772-10G A112772-1G A112772-5G A112772-25G
103798-100EA 9 USD
103798-100 103823-938 76765-982 76765-984 77310-758 77310-760 77623-930
6-Methyl-2-pyridinemethanol 97%
6-Methyl-2-pyridinemethanol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.16 g/mol
Flash Pt: 108.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00023520
CAS Number: 1122-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR