6-Methylpicolinic acid 98%
Supplier: Ambeed
|
Warning
|
A137131-100G
A137131-10G
A137131-1G
A137131-25G
A137131-5G
103802-416EA
6
USD
103802-416
76871-000
76871-002
77315-370
77315-372
77315-374
6-Methylpicolinic acid 98%
6-Methylpicolinic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₂ Melting Pt: 120…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023481 CAS Number: 934-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...