2-Formyl-6-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde

A143143-100G A143143-10G A143143-1G A143143-25G A143143-5G
103803-244EA 6 USD
103803-244 77137-420 77137-422 77316-490 77316-492 77316-494
2-Formyl-6-picoline 97%
2-Formyl-6-picoline

Precaución: for Research Use Only

Formula: C₇H₇NO
MW: 121.14 g/mol
Melting Pt: 31…33 °C
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006291
CAS Number: 1122-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR