6-Methyl-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

A116169-100MG A116169-250MG A116169-1G A116169-5G
103798-686EA 40 USD
103798-686 103824-550 76766-202 76766-204 77297-430 77297-432
6-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
6-Methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
Melting Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659664
CAS Number: 824-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR