6-Methyl-1H-indole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A243469-1G A243469-250MG
103811-518EA 13 USD
103811-518 103851-340 77090-534 77626-298
6-Methyl-1H-indole-3-carboxylic acid 95%
6-Methyl-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
Flash Pt: 202.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06203633
CAS Number: 209920-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR