6-Methoxyquinoxalin-2(1H)-one 95%

Supplier: Ambeed
Warning

A326390-100MG A326390-250MG
103825-434EA 343 USD
103825-434 76875-942 76875-944
6-Methoxyquinoxalin-2(1H)-one 95%
6-Methoxyquinoxalin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17215509
CAS Number: 91192-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR