Methyl-6-quinolyl ether 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxyquinoline

A311441-25G A311441-5G A311441-100G A311441-10G A311441-1G
103835-854EA 11 USD
103835-854 76838-648 76838-650 77277-538 77277-540 77277-542
Methyl-6-quinolyl ether 97%
Methyl-6-quinolyl ether

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 295.3 °C (760 mm Hg)
Melting Pt: 18…20 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006800
CAS Number: 5263-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR