6-Methoxy-4-hydroxyquinoline 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxyquinolin-4-ol

A281546-250MG A281546-1G A281546-25G A281546-5G
103813-560EA 12 USD
103813-560 103853-510 76802-202 77305-402 77305-404 77305-406
6-Methoxy-4-hydroxyquinoline 97%
6-Methoxy-4-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Boiling Pt: 351.8 °C (760 mm Hg)
Melting Pt: 241…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00014662
CAS Number: 23432-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR