6-Methoxypyrimidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A230922-100MG A230922-250MG A230922-1G A230922-5G
103810-736EA 135 USD
103810-736 103850-482 76874-238 76874-240 77303-892 77303-894
6-Methoxypyrimidine-4-carboxylic acid 98%
6-Methoxypyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00496784
CAS Number: 38214-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR