6-Methoxypyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A124442-250MG A124442-1G A124442-5G
103800-532EA 100 USD
103800-532 77258-192 77297-990 77297-992
6-Methoxypyrimidin-4(3H)-one 97%
6-Methoxypyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00955675
CAS Number: 6128-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR