6-Methoxypyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

A273049-10G A273049-250MG A273049-1G A273049-5G
103813-122EA 24 USD
103813-122 103853-038 76772-174 76772-176 77092-832 77305-118
6-Methoxypyridine-2-carbonitrile 98%
6-Methoxypyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.135 g/mol
Melting Pt: 62…67 °C
Storage Temperature: Ambient
MDL Number: MFCD08062856
CAS Number: 83621-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR