6-Methoxy-1-naphthol 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxynaphthalen-1-ol

A147613-100MG A147613-250MG A147613-25G A147613-1G A147613-5G
103803-808EA 17 USD
103803-808 103841-914 76798-504 76798-506 76798-508 77317-304 77317-306
6-Methoxy-1-naphthol 98%
6-Methoxy-1-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MW: 174.196 g/mol
Boiling Pt: 336.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03695468
CAS Number: 22604-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR