6-Methoxybenzo[d]thiazole-2(3H)-thione 97%

Supplier: Ambeed
Warning

A425386-25G A425386-5G A425386-250MG
103827-460EA 14 USD
103827-460 103838-510 76842-164 76842-166 77884-097
6-Methoxybenzo[d]thiazole-2(3H)-thione 97%
6-Methoxybenzo[d]thiazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₈H₇NOS₂
Boiling Pt: 348.9 °C (760 mm Hg)
Flash Pt: 164.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22003314
CAS Number: 2182-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR