6-Methoxy-1-benzothiophene 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxybenzo[b]thiophene

A124193-100MG A124193-5G A124193-1G A124193-250MG
103800-498EA 104 USD
103800-498 76870-418 76870-420 77312-630 77670-762
6-Methoxy-1-benzothiophene 98%
6-Methoxy-1-benzothiophene

Precaución: for Research Use Only

Formula: C₉H₈OS
MW: 164.224 g/mol
Boiling Pt: 268.874 °C (760 mm Hg)
Flash Pt: 116.411 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13181215
CAS Number: 90560-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR