6-Methoxy-5-nitropicolinic acid 97%

Supplier: Ambeed
Warning

A575021-5G A575021-100MG A575021-250MG
103828-828EA 148 USD
103828-828 103839-972 76844-104 76991-846 76991-848
6-Methoxy-5-nitropicolinic acid 97%
6-Methoxy-5-nitropicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₅
MW: 198.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18261308
CAS Number: 475272-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR