5-Amino-2-methoxy-4-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-4-methylpyridin-3-amine, 5-Amino-2-methoxy-4-methylpyridine

A117721-250MG A117721-25G
103798-930EA 11 USD
103798-930 103824-820 77108-350 77108-352
5-Amino-2-methoxy-4-picoline 98%
5-Amino-2-methoxy-4-picoline

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.1700 g/mol
Boiling Pt: 280.6 °C (760 mm Hg)
Melting Pt: 157…161 °C
Flash Pt: 124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08277274
CAS Number: 6635-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR