2-Amino-6-methoxy-3-nitropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-3-nitropyridin-2-amine

A163655-100G A163655-25G A163655-5G
103844-002EA 11 USD
103844-002 77110-762 77300-664 77300-666
2-Amino-6-methoxy-3-nitropyridine 97%
2-Amino-6-methoxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₃
MW: 169.14 g/mol
Melting Pt: 170…171 °C
Storage Temperature: Ambient
MDL Number: MFCD03206481
CAS Number: 73896-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR