6-Methoxy-3,4-dihydro-1H-quinolin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-3,4-dihydroquinolin-2(1H)-one

A383882-1G A383882-5G A383882-100MG A383882-250MG
103826-754EA 22 USD
103826-754 103837-790 76841-132 76841-134 76978-854 76978-856
6-Methoxy-3,4-dihydro-1H-quinolin-2-one 97%
6-Methoxy-3,4-dihydro-1H-quinolin-2-one

Precaución: For research use only

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179395
CAS Number: 54197-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR