6-Methoxy-1-tetralone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-1,2,3,4-tetrahydro-1-naphthalenone, 6-Methoxy-3,4-dihydronaphthalen-1(2H)-one

A156034-100G A156034-10G A156034-500G A156034-5G A156034-25G
103843-024EA 5 USD
103843-024 76798-798 76798-800 76798-802 77300-158 77318-766
6-Methoxy-1-tetralone 98%
6-Methoxy-1-tetralone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 171 °C (760 mm Hg)
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001695
CAS Number: 1078-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR