6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A174169-10G A174169-250MG A174169-1G A174169-25G A174169-5G
103806-928EA 36 USD
103806-928 103845-226 77011-700 77011-702 77082-516 77082-518 77082-520
6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one 97%
6-Methoxy-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
Boiling Pt: 439.8 °C (760 mm Hg)
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD04114865
CAS Number: 22246-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR