6-Methoxy-2-pyrazinecarboxylic acid 98%

Supplier: Ambeed
Warning

A412001-1G A412001-100MG A412001-250MG A412001-5G
103827-236EA 43 USD
103827-236 76841-824 76979-156 76979-158 76979-160
6-Methoxy-2-pyrazinecarboxylic acid 98%
6-Methoxy-2-pyrazinecarboxylic acid

Precaución: For research use only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 176…181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233277
CAS Number: 24005-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR